DNA gel loading buffer 6X
Lieferant: LONZA
50655
LONZ50655EA
75.3
EUR
LONZ50655
DNA gel loading buffer 6X
Puffer
Elektrophorese-Puffer
Beladungspuffer
This buffer is a Ficoll based loading buffer containing Ficoll®, EDTA, xylene cyanol, and bromophenol blue. Ideal for separation of DNA fragments with improved resolution.