DNA gel loading buffer 6X
Lieferant: LONZA
50655
LONZ50655EA 75.3 EUR
LONZ50655
DNA gel loading buffer 6X
Puffer Elektrophorese-Puffer Beladungspuffer
This buffer is a Ficoll based loading buffer containing Ficoll®, EDTA, xylene cyanol, and bromophenol blue. Ideal for separation of DNA fragments with improved resolution.