Natriumpyruvat-Lösung 100 mM Reagenz für die Zellkultur

Lieferant: LONZA

Synonyme: 2-Oxopropionsäure Natriumsalz, Brenztraubensäure Natriumsalz, 2-Ketopropionsäure Natriumsalz, α-Ketopropionsäure Natriumsalz

BEBP13-115E
LONZBEBP13-115EEA 31.8 EUR
LONZBEBP13-115E
Natriumpyruvat-Lösung 100 mM Reagenz für die Zellkultur
Natriumpyruvat

Sodium Pyruvate Solution is sometimes added to cell culture as an additional source of energy since it is a key intermediate in the production of the high energy intermediate ATP within the cell. Its normal concentration in MEM is 1 mM.


Sodium Pyruvate Solution may help cells that are deficient in the nine enzymatic steps that break glucose down to pyruvate.
Sodium Pyruvate Solution may also assist in “jump starting” cells in the citric acid cycle and thus help low density cells grow and produce energy in the form of ATP.


All BioWhittakerTM Cell Culture Media products are for Research Use Only (RUO) and are not approved for human or veterinary use or for use in clinical or in vitro diagnostic procedures

Formel: CH₃COCOONa
Molecular Weight: 110,04 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00002586
CAS-Nummer: 113-24-6
EINECS: 204-024-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR