Staining jars acc.to Hellendahl
Lieferant: LP ITALIANA
290417
LPIT290417EA 79.4 EUR
LPIT290417
Staining jars acc.to Hellendahl
Dosen Färbeküvetten
Jar made of TPX.
It hold up to 16 microscopic slides.