Methylblau, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Baumwollblau, Acid Blue 93, Anilinblau WS, Wasserblau 6B, Aniline blue water soluble, Anilinblau wasserlöslich, Aniline blue WS, Dinatrium-[[4-[bis[4-[(sulfonatophenyl)amino]phenyl]methylen]cyclohexa-2,5-dien-1-yliden]amino]benzolsulfonat

M-Clarity™ Qualitätsstufe = MQ200

M5528-25G
M5528-25GEA 195 EUR
M5528-25G
Methylblau, Sigma-Aldrich®
Methylblau
Formel: C₃₇H₂₇N₃Na₂O₉S₃
Molecular Weight: 799,81 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058509
CAS-Nummer: 28983-56-4
EINECS: 249-352-9
Merck Index: 13,06055
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0dd93fd8c850acc25ffe6d5612e577a0fc82e06&vid=bb467e879ac96a695c323c92253bf4d989856559&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR