TLC education advanced kit, micro-set F3
Lieferant: MACHEREY-NAGEL
814400 814401 814402 814403 814404 814405 814406 814407
MANA814400EA 265 EUR
MANA814400 MANA814401 MANA814402 MANA814403 MANA814404 MANA814405 MANA814406 MANA814407
TLC education advanced kit, micro-set F3
Lehrmittel Chemie-Lehrmittel Chromatographie-Lehrmittel
Advanced kit for science education.
  • All chemicals included for the separation of:
  • Analgetics (pain relievers)
  • Drugs as shown for cinchona bark
Together with the materials from TLC micro-Set M Aspirin® and Thomapyrin® tablets as well as the drug cinchona bark can be separated and visualised by spray reagents (e.g., reagent according to Dragendorff-Munier).
Bitte beachten * These products contain harmful substances which must be specially labelled as hazardous.