Photometric tube tests, NANOCOLOR® cationic Surfactants
Lieferant: MACHEREY-NAGEL
985034
MANA985034EA 112 EUR
MANA985034
Photometric tube tests, NANOCOLOR® cationic Surfactants
Umwelt-Prüfkits
A pre-dosed NANOCOLOR tube test for photometric determination of cationic surfactants in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes