Photometric tube tests, NANOCOLOR® cationic Surfactants
Lieferant: MACHEREY-NAGEL
985034
MANA985034EA
112
EUR
MANA985034
Photometric tube tests, NANOCOLOR® cationic Surfactants
Umwelt-Prüfkits
A pre-dosed NANOCOLOR tube test for photometric determination of cationic surfactants in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (OD) tubes