Microscope slide for LCP
Lieferant: MARIENFELD
0890003
MARI0890003EA 463.6 EUR
MARI0890003
Microscope slide for LCP
Objektträger Mikroskop-Objektträger
The lipidic cubic phase (LCP) sandwich set consists of a base slide and a cover slip and is optimized for the crystallographic study of membrane proteins.
  • Surface with hydrophobic glass
  • Optimised for brightfield, UV and fluorescent microscopy
  • Can be used in G protein–coupled receptors (GPCR) structural biology