Sicherheitsmesser, SECUNORM PROFI25 MDP
Lieferant: MARTOR ARGENTAX
120700.02
MARO120700.02EA 37.1 EUR
MARO120700.02
Sicherheitsmesser, SECUNORM PROFI25 MDP
Messer
Trapezklinge, Griff aus Aluminium.
  • With spring loaded blade retraction, designed to respond to any uncontrolled move if and when the blade slider is not in forward position
  • Can be used by left and right-handed users
  • Easy blade change: No extra tools needed
  • Stainless 'food contact approved' blades
This safety cutter with aluminium body with metal-detectable blue plastic parts and stainless 'food contact approved' blades is ideal for slitting cardboard and adhesive tape.
TÜV tested
Standards: TÜV-geprüft.