1-(2-Chlor-6-fluorbenzyl)piperazin

Lieferant: Thermo Scientific

AW00532.250
MAYBAW00532.250EA 0 EUR
MAYBAW00532.250
1-(2-Chlor-6-fluorbenzyl)piperazin
1-(2-Chlor-6-fluorbenzyl)piperazin
Formel: C₁₁H₁₄ClFN₂
Molecular Weight: 228,7 g/mol
MDL: MFCD00816605
CAS-Nummer: 215655-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR