Paraffin, Plätzchen, MEDITE PURE™
Synonyme:
Wax Paraffin, Paraffin wax
40-0021-00
MEDT40-0021-00EA
105
EUR
MEDT40-0021-00
Paraffin, Plätzchen, MEDITE PURE™
Paraffin
CAS-Nummer:
8002-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...