Biopsy punch with plunger system
Lieferant: MEGRO
KAI-P101
MEGRKAI-P101EA 175 EUR
MEGRKAI-P101
Biopsy punch with plunger system
Biopsiestanzen
The integrated collection aid (plunger system) permits the fast and easy removal of the skin sample from the biopsy punch.
  • Ribbed handle
  • Stainless steel blade