Oven Sample Processor, 874
Lieferant: METROHM
28740120
METR28740120EA 0 EUR
METR28740120
Oven Sample Processor, 874
Instrumente zur Probenvorbereitung
The 874 Oven Sample Processor is used for automatic thermal sample preparation in Karl Fischer titration.
  • Oven sample changer: Automated sample preparation for samples that cannot be titrated directly
The oven method is particularly suitable for samples that do not release their water until higher temperatures have been reached, for sparingly soluble samples, or those that react with the KF reagent.