Temperature Sensor for CVS, PT1000
Lieferant: METROHM
6.1110.120
METR6.1110.120EA 711 EUR
METR6.1110.120
Temperature Sensor for CVS, PT1000
Temperaturfühler
PT1000 temperature sensor (class B) for professional CVS instruments.
  • Shaft diameter top: 12 mm
  • Shaft diameter bottom: 5 mm
  • Minimum immersion depth: 20 mm