Conductivity standards
Lieferant: Mettler - Toledo
51350094
METT51350094EA
29.5
EUR
METT51350094
Conductivity standards
Standards
Leitfähigkeits-Standards
A slight change in temperature usually has a big impact on the conductivity value of a standard solution. A table on every bottle label indicates the conductivity values at the most common measurement temperatures.
During calibration the meter automatically references to this table for temperature compensation. If possible, calibration and measurements should be performed at the same temperature