Masterflex® Polypropylene Felt Filter Bags, Avantor®
Lieferant: Avantor Fluid Handling
01519-72
MFLX01519-72EA 5.5 EUR
MFLX01519-72
Masterflex® Polypropylene Felt Filter Bags, Avantor®
Filter Bag Filter Systems
Attach bags onto any open system drain or sump-return to collect solids.
  • Solids are contained within the bag for easy disposal
An improvement over tie-on designs—the head and support basket make the full filter bag surface area available for use. Filter bags include a galvanized steel ring sewn into the top that securely holds the bag to the head. Polypropylene bags are glazed to reduce fiber migration and have welded seams for high strength. All bag designs are suitable to 100 °F (38 °C).