ProtEX-RTs Explosion-Proof Rate/Totalizers
Lieferant: Avantor Fluid Handling
30004-63
MFLX30004-63EA 994 EUR
MFLX30004-63
ProtEX-RTs Explosion-Proof Rate/Totalizers
Überwachungssysteme Prozessüberwachung
Program your process meter on-site.
  • 5-digit rate and 7-character total/tag display
  • Backlight standard
  • Rotatable display
  • FM, CSA, ATEX, IECEx, CE approved
The ProtEX-RTs are rugged, explosion-proof, full-featured rate/totalizers designed for demanding applications in hazardous areas or in the harshest environmental conditions. The PD6820 accepts analog inputs and the PD6830 accepts pulse inputs. They are programmed using the four SafeTouch™ through-glass buttons without removing the cover, or the four mechanical buttons. The sleek die-cast aluminum enclosure is provided with two ¾-inch threaded conduit holes and integrated pipe or wall mounting flanges.