Miniature Plastic Hose Barb Check Valves
Lieferant: Avantor Fluid Handling
98553-02
MFLX98553-02EA 7.2 EUR
MFLX98553-02
Miniature Plastic Hose Barb Check Valves
Ventile Rückschlagventile
Use with liquids and gases for pressure or vacuum applications.
  • Breaking pressure is 0.18 psi, closing pressure is 0.014 psi
  • Install in any position
Note: Valves require 1 psi back pressure for a tight seal.