Hämatoxylin
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
DcH-48, Natural Black I, Hydroxybrasilin
M-Clarity™ Qualitätsstufe = MQ500
MHS80-2.5L
MHS80-2.5LEA
209
EUR
MHS80-2.5L
Hämatoxylin
Haematoxylin staining solutions
Formel:
C₁₆H₁₄O₆ Molecular Weight: 302,28 g/mol |
MDL:
MFCD00078111 CAS-Nummer: 517-28-2 |
Spezifikation Testergebnisse
Appearance (Colour) | Red-Brown |
Appearance (Form) | Solution |
Appearance (Turbidity) | Clear |
Performance | Conforms |
Stains within acceptable limits when using Procedure MHS | |
Expiration Date Period | 36 Months |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...