Plastic Tube Brushes
Lieferant: Micronova
MICRBRPL-3418EA 58.7 EUR
MICRBRPL-3418 MICRBRPL-25S36 MICRBRPL-1218 MICRBRPL-1418 MICRBRPL-118
Plastic Tube Brushes
Bürsten
These durable, 100% polypropylene brushes clean finely polished stainless steel parts without scratching.
  • 100% polypropylene bristles and handle
  • Thin core and handles are flexible enough to bend through fittings
  • Available in a range of bristle diameters and handle lengths
Brushes feature narrow center cores that are flexible enough to pass through elbow tank fittings.
Standards: FDA compliant materials.