Filter forceps
Lieferant: Merck
XX6200006P
MILFXX6200006PEA 76.7 EUR
MILFXX6200006P
Filter forceps
Pinzetten
Sterilisable filter forceps made from stainless steel, with PP covered handles (in three different colours).