Flask and Neoprene Stopper
Lieferant: Merck
XX6602501
MILFXX6602501EA 449 EUR
MILFXX6602501
Flask and Neoprene Stopper
Kolben Saugflaschen
Simultaneously vacuum filter 12 samples for rapid analysis of retentate or filtrate. Numbered positions on the filter plate assure accurate sample tracking. Sealing stoppers maintain vacuum on unused sample cups. Accessory extension barrels increase sample volumes from 15 to 50 ml.
General filtration of 15 to 50 ml samples, preparation for scintillation counting.