Protein ladders (pre-stained), ScanLater™
Lieferant: Molecular Devices
R8220
MLDVR8220EA 331 EUR
MLDVR8220
Protein ladders (pre-stained), ScanLater™
Elektrophorese-Reagenzien Molekulargewichtsmarker
The ScanLater™ Western blot protein ladder is a mixture of seven biotinylated recombinant proteins at 10, 20, 40, 60, 80, 100, and 140 kD.
  • 3 pre-stained marker proteins at 18, 31, and 70 kD
  • Ready to use, for 30 mini gels with recommended protocol