DBI-SALA® Safe Bucket
Lieferant: 3M
1500134
MMMA1500134EA 196 EUR
MMMA1500134
DBI-SALA® Safe Bucket
Eimer
3M™ DBI SALA® Safe Bucket is designed to be the perfect companion for workers at height carrying heavy loads. It is manufactured with either a hook and loop or drawstring closure system which helps prevent accidental drops. Colorful indicator tabs provide quick visual cues to indicate whether the bucket is in lockdown mode or easy-access mode.
  • Compatible with 3M™ DBI-SALA® and Protecta® Fall Protection systems and components
  • Enable workers to secure tools and accessories while working at height
  • Multiple materials, closure systems, and attachment point options designed for a variety of applications
Built-in connection points, the Safe Bucket was engineered for use with tools at height, and includes six connection points for tethering. Available in both heavy-duty canvas and vinyl variants that are load rated for 45,4 and 91 kg respectively.
Weight: 1,83 kg
Weight capacity: 45,4 kg
Overall length: 448,31 mm
Recommended industry: Construction, general industrial, mining, oil and gas, transportation.