3M™ Combination Filter
Lieferant: 3M
7100260645
MMMA7100260645EA 39.7 EUR
MMMA7100260645
3M™ Combination Filter
Respirators Mehrweg-Atemschutzmasken Maskenfilter
For effective protection against a range of particulates, gases and vapours or combination hazards, use 3M™ Filters DT-Series. Suitable for use with certain 3M™ half and full-face tight-fitting respirators in negative-pressure mode as well as 3M™ Powered Air Respirator System PF-600E+ Series.
  • Includes specifications to provide protection against particulate hazards, gas & vapour hazards or a combination of both
  • Features a DIN40 mm threaded connection
  • Please note DT-Series filters were previously known as Pro2000 Filters
  • Suitable for use with 3M™ Powered Air Respirators PF-602E+ and PF-602E-ASB. Note: A1 classification when used with powered air systems
Protection type: Acid gas, ammonia, iorganic vapor, mercury, multi-gas, organic vapour, particulate
Recommended application: Demolition, grinding, metalworking