Nomad™ 4300 Ultra Clean Mattings
Lieferant: 3M
ADHBU459
MMMAADHBU459EA 771 EUR
MMMAADHBU459
Nomad™ 4300 Ultra Clean Mattings
Fußmatten Klebematten
Nomad™ ultra clean matting 4300 is a disposable, dust collecting floor mat consisting of 40 sheets coated with a pressure sensitive adhesive.
  • Disposable multi-layered contamination control mat
  • Peel off one layer to expose the next
  • Lay onto any clean surface with no edging or holding frame required
  • Anti-microbial additive stops the growth of micro-organisms
High tack surface removes dust and small particles from shoe soles and trolley wheels. When the top layer is contaminated, it is then simply peeled off to expose the next fresh adhesive layer ready for immediate use.
The mat can be laid directly onto any clean floor surface and the low profile design does not need any edging or frame. It is designed to help protect clean areas where cleanliness and hygiene are critical factors.