PELTOR™ X5 Earmuff, Helmet Mounted
Lieferant: 3M
X5P5E
MMMAX5P5EEA 64 EUR
MMMAX5P5E
PELTOR™ X5 Earmuff, Helmet Mounted
Kapselgehörschützer Lärmschutz-Kapselgehörschützer
3M™ PELTOR™ X5 Earmuffs are innovative hearing protectors that offer exceptional performance, without the need for double protection. Offering the same premium features as the 3M™ PELTOR™ X Series, they also reduce noise up to 37 dB.
  • Reducing noise levels from 36 to 37 dB, model dependant, tested to EN 352:2020
  • Available as electrically insulated and resistant, or 'dielectric' in A band and P5 helmet attached models
  • Twin headband design provides heat dispersion
  • Internal stainless steel design offers consistent pressure and comfortable fit over an 8 hour period
  • Designed to optimise weight distribution and balance for outstanding comfort
  • ABS plastic cups are rigid, high-strength, and impact-resistant
  • Thermoplastic polyurethane (TPU) soft outer cup coating protects from abrasion and bumps
For high performance hearing protection against moderate to high noise levels, use 3M™ PELTOR™ X5 Earmuffs. The X5 earmuffs are colour-coded black, making it simple to select the correct level of protection from the X series range. These earmuffs feature a specially formulated foam in the earcups, and cushions with an innovative spacer and cup design. These are the highest attenuating earmuffs in the PELTOR™ range, reducing noise up to 37 dB. The technology employed in the design means despite the larger earcups, the earmuff is still lightweight with excellent balance and comfort.
The headband model features an electrically insulated, twin headband design that helps reduce heat build-up on top of the head. The stainless steel wire, coated with plastic, makes the headband the most robust 3M™ has to offer. It is both strong and comfortable as the flat wide headband has a soft plastic coating providing stable pressure on top of the head. No exposed metal means the product is 'dielectric' providing electrical resistance for those applications with risk of spark or charge. Carefully moulded sliders ensure the earmuffs are easy to adjust when on the head. Wide, soft foam ear cushions help workers stay comfortable, while the slim cups offer improved compatibility with other safety equipment.
The helmet mounted version is available as P5 with the same dielectric performance as the headband model. Alternatively, if the application does not require electrical insulation the helmet mounted P3 model and neckband is available.
The cushions and foam inserts, available as Hygiene Kits, are easy to replace if they become worn or damaged, helping extend the life of the product.