Double channel thermometers
Lieferant: MOINEAU
2041T
MOIN2041TEA 61.1 EUR
MOIN2041T
Double channel thermometers
Thermometer Digitalthermometer
These 2-channel thermometers feature an electronic digital LCD with dual display of internal and extrenal temperatures.
  • Self-standing or wall mountable
  • Programmable high and low alarms (model 2041T only)
  • Max./Min. memory
  • External sensor according to IP67 (model 2906TM only)
External sensors feature a 1 m cable.