Digital timer, three channels
Lieferant: MOINEAU
419CA
MOIN419CAEA 59.6 EUR
MOIN419CA
Digital timer, three channels
Timer Timer, digital
Three-channel (1× count up, 2× count down) timer with metal housing.
  • Timing capacity: 24 hours
  • Two alarms, two memories, snooze function
  • Clock (AM/PM)
Lieferumfang: Supplied with stand, clip, magnet and LR44 battery.