Earmuffs, foldable, M1
Lieferant: Moldex
6100
MOLD6100EA 49.8 EUR
MOLD6100
Earmuffs, foldable, M1
Kapselgehörschützer Lärmschutz-Kapselgehörschützer
M1 Premium foldable earmuff with exclusive metallic colour and SoftCoat® covering that helps muffle transmitted noise.
  • Features a black-chrome, spring-steel padded headband that resists fatigue and is more durable
  • Maintains stable and does not twist out of shape
  • Exclusive iridescent colour and curvy shape for great looks
  • Six different adjustment points provide a custom and comfortable fit for most head sizes
  • Folds into compact size for easier storage and hygiene
SNR: 33 dB
Standards: EN 352-1:2002