Hydrochlorothiazid

Lieferant: Molekula

10985573-25G
MOLE10985573-25GEA 92.1 EUR
MOLE10985573-25G
Hydrochlorothiazid
Hydrochlorothiazid
Formel: C₇H₈ClN₃O₄S₂
MDL: MFCD00051765
CAS-Nummer: 58-93-5
EINECS: 200-403-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR