L(-)-Arabitol
Lieferant: Molekula
Synonyme:
L-(-)-Arabinitol, (2S,4S)-Pentane-1,2,3,4,5-pentaol, L-(-)-Arabitol, L-Arabinitol
25675490-25G
MOLE25675490-25GEA
219
EUR
MOLE25675490-25G
L(-)-Arabitol
L(-)-Arabitol
Formel:
C₅H₁₂O₅ Molecular Weight: 152,15 g/mol |
MDL:
MFCD00064290 CAS-Nummer: 7643-75-6 EINECS: 231-582-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...