Uridin
Lieferant: Molekula
Synonyme:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
26362922-100G
MOLE26362922-100GEA
144
EUR
MOLE26362922-100G
Uridin
Uridin
Formel:
C₉H₁₂N₂O₆ Molecular Weight: 244,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006526 CAS-Nummer: 58-96-8 EINECS: 200-407-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...