Tetradecyltrimethylammoniumchlorid

Lieferant: Molekula

Synonyme: Myristyltrimethylammoniumchlorid, Tetradoniumchlorid, TTAC

28986024-25G
MOLE28986024-25GEA 0 EUR
MOLE28986024-25G
Tetradecyltrimethylammoniumchlorid
Tetradecyltrimethylammoniumchlorid
Formel: C₁₇H₃₈ClN
Molecular Weight: 291,95 g/mol
MDL: MFCD00059968
CAS-Nummer: 4574-04-3
EINECS: 224-958-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR