L(+)-Cystein Hydrochlorid

Lieferant: Molekula

Synonyme: H-Cys-OH.HCl, (R)-(+)-Cystein Hydrochlorid, (R)-(+)-2-Amino-3-mercaptopropansäure Hydrochlorid

29268471-1KG
MOLE29268471-1KGEA 360 EUR
MOLE29268471-1KG
L(+)-Cystein Hydrochlorid
L(+)-Cystein Hydrochlorid
Formel: HSCH₂CH(NH₂)COOH·HCl
Molecular Weight: 157,62 g/mol
Schmelzpunkt: 175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064553
CAS-Nummer: 52-89-1
EINECS: 200-157-7
Merck Index: 13,02810

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR