Ponceau S
Lieferant: Molekula
|
Achtung
|
Synonyme:
3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
43797916-25G
MOLE43797916-25GEA
134
EUR
MOLE43797916-25G
Ponceau S
Ponceau S
Formel:
C₂₂H₁₂N₄Na₄O₁₃S₄ Molecular Weight: 760,56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003892 CAS-Nummer: 6226-79-5 EINECS: 228-319-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...