5-Bromisochinolin

Lieferant: Molekula
Achtung

48905952-1G
MOLE48905952-1GEA 0 EUR
MOLE48905952-1G
5-Bromisochinolin
5-Bromoisoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD01646405
CAS-Nummer: 34784-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR