Alcianblau 8GX

Lieferant: Molekula
Achtung

59508864-10G
MOLE59508864-10GEA 142 EUR
MOLE59508864-10G
Alcianblau 8GX
Alcian blue 8 GX
Formel: C₅₆H₆₈Cl₄CuN₁₆S₄
Molecular Weight: 1298,86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010720
CAS-Nummer: 33864-99-2
EINECS: 251-705-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR