Mitomycin C

Lieferant: Molekula
Gefahr

Synonyme: Mitomycin, Ametycin

M20540412
MOLEM20540412EA 87.3 EUR
MOLEM20540412
Mitomycin C
Mitomycin C
Formel: C₁₅H₁₈N₄O₅
Molecular Weight: 334,33 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00078109
CAS-Nummer: 50-07-7
EINECS: 200-008-6
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR