2',7'-Dichlorfluorescein

Lieferant: Molekula

Synonyme: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

M27741374
MOLEM27741374EA 0 EUR
MOLEM27741374
2',7'-Dichlorfluorescein
2',7'-Dichlorfluorescein
Formel: C₂₀H₁₀Cl₂O₅
Molecular Weight: 401,20 g/mol
MDL: MFCD00005047
CAS-Nummer: 76-54-0
EINECS: 200-968-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR