Tetracyclin Hydrochlorid

Lieferant: Molekula
Achtung

M44594076
MOLEM44594076EA 0 EUR
MOLEM44594076
Tetracyclin Hydrochlorid
Tetracyclin Hydrochlorid
Formel: C₂₂H₂₅ClN₂O₈
Molecular Weight: 480,9 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078142
CAS-Nummer: 64-75-5
EINECS: 200-593-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR