Tetrabutylammoniumperchlorat
Lieferant: Molekula
Synonyme:
Tetra-n-butylammonium perchlorate
M62084577
MOLEM62084577EA
0
EUR
MOLEM62084577
Tetrabutylammoniumperchlorat
Tetrabutylammonium perchlorate
Formel:
C₁₆H₃₆ClNO₄ Molecular Weight: 341,92 g/mol Schmelzpunkt: 210 °C Dichte: 1,07 g/cm³ (20 °C) |
MDL:
MFCD00038722 CAS-Nummer: 1923-70-2 EINECS: 217-655-5 UN: 1481 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...