3-Hydroxypicolinsäure

Lieferant: Molekula

Synonyme: 3-Hydroxypyridine-2-carboxylic acid

M84867525
MOLEM84867525EA 0 EUR
MOLEM84867525
3-Hydroxypicolinsäure
3-Hydroxypicolinsäure
Formel: C₆H₅NO₃
Molecular Weight: 139,11 g/mol
MDL: MFCD00006294
CAS-Nummer: 874-24-8
EINECS: 212-859-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR