Safety PE Storage Can with Dosing Tap
Lieferant: asecos
40096
MSGA40096EA 91 EUR
MSGA40096
Safety PE Storage Can with Dosing Tap
Sicherheitsbehälter
The PE Storage Can is ideal for the safe storage and internal transport of liquids, such as acids or lyes.
  • With fine dosing tap made of stainless steel (1.4301), includes vent, PTFE seal
  • Includes multilingual safety sticker to designate the contents