1,3-Propandiol
Lieferant: MARKET SOURCE PART PROCESS
Synonyme:
Trimethylenglycol, 1,3-Propylenglycol, 1,3-Dihydroxypropan
5495260000
MSPP5495260000EA
0
EUR
MSPP5495260000
1,3-Propandiol
1,3-Propandiol
Formel:
HO(CH₂)₃OH Molecular Weight: 76,1 g/mol Siedepunkt: 214 °C (1013 hPa) Schmelzpunkt: –32 °C Dichte: 1,0597 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002949 CAS-Nummer: 504-63-2 EINECS: 207-997-3 Merck Index: 12,09845 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...