Quick-DNA™ Fungal/Bacterial Kits
Lieferant: MARKET SOURCE PART PROCESS
Quick, efficient isolation of DNA from all types of tough-to-lyse fungi and bacteria.
- Simple, efficient isolation of DNA from all types of tough-to-lyse fungi and bacteria in minutes
- Ultra-high density BashingBeads™ are fracture resistant and chemically inert
The Quick-DNA™ Fungal/Bacterial Kits are designed for the simple and rapid isolation of DNA from tough-to-lyse fungi, including A. fumigatus, C. albicans, N. crassa, S. cerevisiae, S. pombe, as well as Gram-positive/negative bacteria, algae, and protozoa. The procedures are easy and can be completed in minutes. Fungal and/or bacterial samples are rapidly and efficiently lysed with our state-of-the-art, ultra-high density BashingBeads™. Zymo-Spin™ technology is then used to isolate the DNA, which is ideal for downstream molecular-based applications including PCR, array and more.