Zellulose

Lieferant: MARKET SOURCE PART PROCESS

Synonyme: Zellulose

PH-200NF
MSPPPH-200NFEA 0 EUR
MSPPPH-200NF
Zellulose
Zellulose
Formel: (C₁₂H₂₂O₁₁)n
Molecular Weight: 342,3 g/mol
Schmelzpunkt: 260…270 °C
Dichte: 1,27…1.6 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00081512
CAS-Nummer: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR