(3-(Dimethylamino)propyl)trimethoxysilan

Lieferant: MARKET SOURCE PART PROCESS

Synonyme: Trimethoxy(3-(dimethylamino)propyl)silan, N,N-Dimethyl-3-(trimethoxysilyl)propylamin, (3-Dimethylaminopropyl)trimethoxysilan

SID3546.94-10G
MSPPSID3546.94-10GEA 0 EUR
MSPPSID3546.94-10G
(3-(Dimethylamino)propyl)trimethoxysilan
(3-(Dimethylamino)propyl)trimethoxysilan
Formel: (H₃C)₂NCH₂CH₂CH₂Si(OCH₃)₃
Molecular Weight: 207,35 g/mol
MDL: MFCD00053674
CAS-Nummer: 2530-86-1
EINECS: 219-786-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR