Zellulose
Lieferant: MARKET SOURCE PART PROCESS
Synonyme:
Zellulose
VIVAPUR102
MSPPVIVAPUR102EA
0
EUR
MSPPVIVAPUR102
Zellulose
Zellulose
Formel:
(C₁₂H₂₂O₁₁)n Molecular Weight: 342,3 g/mol Schmelzpunkt: 260…270 °C Dichte: 1,27…1.6 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081512 CAS-Nummer: 9004-34-6 EINECS: 232-674-9 Merck Index: 13,01977 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...