Screw cap, without dropper
Lieferant: MUELLER KREMPEL
14.016.19
MUEL14.016.19EA 27.8 EUR
MUEL14.016.19
Screw cap, without dropper
Verschlüsse Flaschenverschlüsse
Self-sealing caps without drip insert, suitable for steam sterilization.