Euro-line Hinged Lid
Lieferant: MULTIROIR
BECC64
MULCBECC64EA 49.7 EUR
MULCBECC64
Euro-line Hinged Lid
Deckel Lids for Laboratory Vessels/Containers
Hinged lids allow to secure the lid to the tray. Goods are thus protected during transport or storage.
  • Comes with lid and clasps.
  • Compatible with Euro-line bins
  • Stackable and recyclable
  • Food contact quality
  • Solid walls
The lid is very easily attached to the tank by means of a clipping. Opening and closing are carried out with two blue clasps.
Temperature of use: –20°C / +50°C.